siRNA Details
| Gene Symbol: TMEM132D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133448.2 | NP_597705.2 | 9 | 2595 | ||
| GenBank mRNA | AB061814.1 | - | 9 | 2596 | ||
| AB075824.1 | - | 4 | 2122 | |||
| BC114626.1 | - | 9 | 2595 | 40082027 | ||
| BC114936.1 | - | 9 | 2595 | 40082028 | ||

TMEM132D
Genomic Map