siRNA Details
| Gene Symbol: C12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152319.3 | NP_689532.1 | 2, 3 | 196 | ||
| GenBank mRNA | AY255793.1 | - | 1, 2 | 101 | ||
| AY255794.1 | - | 1, 2 | 101 | |||
| BC031670.2 | - | 1, 2 | 122 | 5165130 | ||
| BQ429029.1 | - | 2, 3 | 199 | |||

C12orf54
Genomic Map