siRNA Details
Gene Symbol: C12orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152319.3 | NP_689532.1 | 2 | 174 | ||
GenBank mRNA | AY255793.1 | - | 1 | 79 | ||
AY255794.1 | - | 1 | 79 | |||
BC031670.2 | - | 1 | 100 | 5165130 | ||
BQ429029.1 | - | 2 | 177 |
