siRNA Details
| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114176.1 | NP_001107648.1 | 1, 2 | 703 | ||
| NM_153207.4 | NP_694939.2 | 1, 2 | 703 | |||
| XM_006719031.3 | XP_006719094.1 | 1, 2 | 885 | |||
| XM_011520560.2 | XP_011518862.1 | 1, 2 | 885 | |||
| GenBank mRNA | AB209384.1 | - | 1, 2 | 497 | ||
| AK126973.1 | - | 2, 3 | 489 | |||
| AK131361.1 | - | 1, 2 | 703 | |||
| BC015624.2 | - | 1, 2 | 190 | 4843866 | ||
| BC022220.1 | - | 1, 2 | 678 | 3914323 | ||

AEBP2
Genomic Map