siRNA Details
| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114176.1 | NP_001107648.1 | 2 | 877 | ||
| NM_001267043.1 | NP_001253972.1 | 2 | 327 | |||
| NM_153207.4 | NP_694939.2 | 2 | 877 | |||
| XM_006719031.3 | XP_006719094.1 | 2 | 1059 | |||
| XM_011520560.2 | XP_011518862.1 | 2 | 1059 | |||
| XM_017018806.1 | XP_016874295.1 | 2 | 204 | |||
| GenBank mRNA | AB209384.1 | - | 2 | 671 | ||
| AK126973.1 | - | 3 | 663 | |||
| AK131361.1 | - | 2 | 877 | |||
| AK131410.1 | - | 2 | 327 | |||
| BC015624.2 | - | 2 | 364 | 4843866 | ||
| BC022220.1 | - | 2 | 852 | 3914323 | ||

AEBP2
Genomic Map