siRNA Details
| Gene Symbol: FOXN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213596.2 | NP_998761.2 | 7 | 766 | ||
| XM_011537922.2 | XP_011536224.1 | 7 | 955 | |||
| XM_011537923.2 | XP_011536225.1 | 7 | 955 | |||
| XM_017018818.1 | XP_016874307.1 | 5 | 1741 | |||
| XM_017018819.1 | XP_016874308.1 | 7 | 955 | |||
| GenBank mRNA | AF425596.1 | - | 5 | 541 | ||
| AF425597.1 | - | 4 | 416 | |||
| AK093286.1 | - | 1 | 44 | |||
| AK131519.1 | - | 3 | 373 | |||
| BC146825.1 | - | 7 | 767 | 8860453 | ||

FOXN4
Genomic Map