siRNA Details
| Gene Symbol: LINC00284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026955.1 | 2, 3 | 413 | |||
| GenBank mRNA | AK001938.1 | - | 1, 2 | 1583 | ||
| AK001941.1 | - | 2, 3 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00284| Gene Symbol: LINC00284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026955.1 | 2, 3 | 413 | |||
| GenBank mRNA | AK001938.1 | - | 1, 2 | 1583 | ||
| AK001941.1 | - | 2, 3 | 260 | |||