siRNA Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145248.4 | NP_660291.2 | 2 | 189 | ||
| XM_011537466.1 | XP_011535768.1 | 2 | 214 | |||
| XM_011537468.2 | XP_011535770.1 | 3 | 275 | |||
| XM_011537471.2 | XP_011535773.1 | 3 | 345 | |||
| XM_011537472.2 | XP_011535774.1 | 2 | 224 | |||
| XM_017020392.1 | XP_016875881.1 | 3 | 290 | |||
| GenBank mRNA | AK130866.1 | - | 2 | 189 | ||
| BC016750.2 | - | 2 | 171 | 4837062 | ||

SPACA7
Genomic Map