siRNA Details
| Gene Symbol: PSMB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099780.1 | NP_001093250.1 | 1 | 638 | ||
| GenBank mRNA | AB299437.1 | - | 1 | 579 | ||
| AK122797.1 | - | 1 | 638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMB11| Gene Symbol: PSMB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099780.1 | NP_001093250.1 | 1 | 638 | ||
| GenBank mRNA | AB299437.1 | - | 1 | 579 | ||
| AK122797.1 | - | 1 | 638 | |||