siRNA Details
| Gene Symbol: KLHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172193.2 | NP_751943.1 | 13 | 1162 | ||
| GenBank mRNA | AF111806.1 | - | 13 | 1100 | ||
| AK127202.1 | - | 13 | 1085 | |||
| AL833437.1 | - | 8 | 592 | |||
| BC031270.1 | - | 13 | 1125 | |||
| BC101595.1 | - | 13 | 1162 | 8069101 | ||
| BC101597.1 | - | 13 | 1162 | 8069103 | ||

KLHDC1
Genomic Map