siRNA Details
| Gene Symbol: C16orf46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100873.1 | NP_001094343.1 | 2 | 324 | ||
| NM_152337.2 | NP_689550.2 | 3 | 413 | |||
| XM_005255792.2 | XP_005255849.1 | 3 | 361 | |||
| XM_011522848.2 | XP_011521150.2 | 3 | 551 | |||
| XM_011522849.2 | XP_011521151.2 | 3 | 580 | |||
| XM_017022916.1 | XP_016878405.1 | 3 | 361 | |||
| XM_017022917.1 | XP_016878406.1 | 2 | 311 | |||
| GenBank mRNA | AK057264.1 | - | 3 | 345 | ||
| BC064143.1 | - | 2 | 270 | 30389327 | ||

C16orf46
Genomic Map