siRNA Details
| Gene Symbol: GSE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023084.1 | XP_016878573.1 | 3 | 4790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSE1| Gene Symbol: GSE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017023084.1 | XP_016878573.1 | 3 | 4790 | ||