siRNA Details
| Gene Symbol: DCUN1D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173475.3 | NP_775746.1 | 3 | 1069 | ||
| GenBank mRNA | AK123719.1 | - | 3 | 1000 | ||
| AY364247.1 | - | 2 | 809 | |||
| BC040442.1 | - | 3 | 1049 | 5590225 | ||
| BX648998.1 | - | 3 | 951 | |||

DCUN1D3
Genomic Map