siRNA Details
| Gene Symbol: IQCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305121.1 | NP_001292050.1 | 5, 6 | 1156 | ||
| NM_001305122.1 | NP_001292051.1 | 6, 7 | 1221 | |||
| NM_153208.2 | NP_694940.1 | 6, 7 | 1221 | |||
| NR_130968.1 | 5, 6 | 1123 | ||||
| XM_011545736.2 | XP_011544038.1 | 5, 6 | 556 | |||
| XM_017022933.1 | XP_016878422.1 | 5, 6 | 1267 | |||
| GenBank mRNA | AF520570.1 | - | 6, 7 | 1221 | ||
| AK301933.1 | - | 5, 6 | 544 | |||
| AK303998.1 | - | 4, 5 | 449 | |||
| AK311216.1 | - | 4, 5 | 416 | |||
| AK315672.1 | - | 5, 6 | 544 | |||
| BC034823.1 | - | 5, 6 | 552 | 5165639 | ||

IQCK
Genomic Map