siRNA Details
| Gene Symbol: ZG16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145252.2 | NP_660295.2 | 4 | 498 | ||
| GenBank mRNA | AK311810.1 | - | 4 | 366 | ||
| AY359021.1 | - | 4 | 440 | |||
| BC009722.1 | - | 4 | 438 | 3879368 | ||
| BM786200.1 | - | 2 | 207 | |||
| BQ924772.1 | - | 4 | 501 | |||
| EF067313.1 | - | 4 | 397 | |||
| JN411265.1 | - | 4 | 439 | |||

ZG16B
Genomic Map