siRNA Details
| Gene Symbol: ZC3H18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294340.1 | NP_001281269.1 | 16 | 2560 | ||
| NM_144604.3 | NP_653205.3 | 15 | 2488 | |||
| XM_011522863.1 | XP_011521165.1 | 14 | 2173 | |||
| XM_011522865.1 | XP_011521167.1 | 8 | 930 | |||
| GenBank mRNA | AK026317.1 | - | 1 | 15 | ||
| AK091849.1 | - | 10 | 1315 | |||
| AK093394.1 | - | 2 | 740 | |||
| AK225807.1 | - | 15 | 2458 | |||
| AK300254.1 | - | 16 | 2538 | |||
| BC001584.1 | - | 12 | 1584 | |||
| BC050463.1 | - | 15 | 2435 | 6042036 | ||

ZC3H18
Genomic Map