siRNA Details
| Gene Symbol: ZC3H18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294340.1 | NP_001281269.1 | 8 | 1460 | ||
| NM_144604.3 | NP_653205.3 | 7 | 1388 | |||
| XM_011522865.1 | XP_011521167.1 | 1 | 99 | |||
| XM_017022934.1 | XP_016878423.1 | 8 | 1460 | |||
| GenBank mRNA | AK091849.1 | - | 2 | 215 | ||
| AK122593.1 | - | 1 | 517 | |||
| AK225807.1 | - | 7 | 1358 | |||
| AK300254.1 | - | 8 | 1438 | |||
| AK302716.1 | - | 7 | 1015 | |||
| BC001584.1 | - | 4 | 484 | |||
| BC050463.1 | - | 7 | 1335 | 6042036 | ||
| DB007291.1 | - | 4 | 474 | |||

ZC3H18
Genomic Map