siRNA Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304996.1 | NP_001291925.1 | 1, 2 | 277 | ||
| NM_145254.2 | NP_660297.1 | 1, 2 | 277 | |||
| GenBank mRNA | AK298475.1 | - | 1, 2 | 229 | ||
| AK311919.1 | - | 1, 2 | 238 | |||
| AY302139.1 | - | 1, 2 | 140 | |||
| BC018082.1 | - | 1, 2 | 226 | 4796761 | ||
| BX647432.1 | - | 1, 2 | 229 | |||
| BX648429.1 | - | 1, 2 | 230 | |||
| BX648484.1 | - | 1, 2 | 230 | |||

TMEM170A
Genomic Map