siRNA Details
| Gene Symbol: CYB5D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144607.4 | NP_653208.2 | 2, 3 | 624 | ||
| XM_005256440.3 | XP_005256497.1 | 2, 3 | 403 | |||
| XM_011523638.2 | XP_011521940.1 | 2, 3 | 403 | |||
| GenBank mRNA | AK057061.1 | - | 2, 3 | 624 | ||
| AK289520.1 | - | 2, 3 | 307 | |||
| BC060779.1 | - | 2, 3 | 376 | 30349502 | ||

CYB5D1
Genomic Map