siRNA Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099225.1 | NP_001092695.1 | 2 | 293 | ||
| NM_144609.2 | NP_653210.2 | 2 | 293 | |||
| GenBank mRNA | AK056357.1 | - | 2 | 258 | ||
| AL833590.1 | - | 2 | 255 | |||
| BC023298.1 | - | 2 | 254 | |||
| BC047776.2 | - | 2 | 259 | 6023964 | ||
| BG706097.1 | - | 2 | 302 | |||
| BX648868.1 | - | 2 | 275 | |||

CCDC43
Genomic Map