siRNA Details
| Gene Symbol: CNTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173478.2 | NP_775749.2 | 2, 3 | 478 | ||
| XM_005257043.4 | XP_005257100.1 | 2, 3 | 241 | |||
| XM_005257044.4 | XP_005257101.1 | 2, 3 | 259 | |||
| XM_006721690.2 | XP_006721753.1 | 2, 3 | 478 | |||
| XM_011524310.2 | XP_011522612.1 | 2, 3 | 425 | |||
| GenBank mRNA | AK097456.1 | - | 2, 3 | 394 | ||
| AK302094.1 | - | 2, 3 | 211 | |||
| BC026187.2 | - | 2, 3 | 522 | 5267540 | ||

CNTD1
Genomic Map