siRNA Details
| Gene Symbol: SPACA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317225.1 | NP_001304154.1 | 4 | 673 | ||
| NM_001317226.1 | NP_001304155.1 | 3 | 296 | |||
| NM_173847.4 | NP_776246.1 | 4 | 605 | |||
| GenBank mRNA | AF216311.1 | - | 4 | 605 | ||
| AY358653.1 | - | 5 | 732 | |||
| BC029867.1 | - | 4 | 673 | 5169651 | ||
| BC100886.2 | - | 4 | 563 | 40003360 | ||
| BC100887.1 | - | 4 | 563 | 40003365 | ||
| BQ438507.1 | - | 3 | 291 | |||

SPACA3
Genomic Map