siRNA Details
| Gene Symbol: SPACA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317225.1 | NP_001304154.1 | 2 | 393 | ||
| NM_173847.4 | NP_776246.1 | 2 | 347 | |||
| GenBank mRNA | AF216311.1 | - | 2 | 347 | ||
| AY358653.1 | - | 3 | 474 | |||
| BC029867.1 | - | 2 | 393 | 5169651 | ||
| BC100886.2 | - | 2 | 305 | 40003360 | ||
| BC100887.1 | - | 2 | 305 | 40003365 | ||

SPACA3
Genomic Map