siRNA Details
| Gene Symbol: TBC1D16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019020.3 | NP_061893.2 | 3 | 800 | ||
| XM_005257049.2 | XP_005257106.1 | 3 | 811 | |||
| XM_005257050.3 | XP_005257107.1 | 3 | 817 | |||
| XM_005257052.3 | XP_005257109.1 | 3 | 817 | |||
| XM_006721694.3 | XP_006721757.1 | 3 | 817 | |||
| XM_006721695.3 | XP_006721758.1 | 3 | 817 | |||
| XM_011524322.2 | XP_011522624.1 | 3 | 817 | |||
| XM_017024185.1 | XP_016879674.1 | 3 | 817 | |||
| GenBank mRNA | AB449902.1 | - | 2 | 674 | ||
| BC028290.1 | - | 3 | 790 | 4480654 | ||

TBC1D16
Genomic Map