siRNA Details
| Gene Symbol: KRT222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152349.2 | NP_689562.1 | 6 | 851 | ||
| GenBank mRNA | BC032815.1 | - | 6 | 810 | 4346009 | |
| BX538083.1 | - | 6 | 804 | |||
| DB543411.1 | - | 1 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT222| Gene Symbol: KRT222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152349.2 | NP_689562.1 | 6 | 851 | ||
| GenBank mRNA | BC032815.1 | - | 6 | 810 | 4346009 | |
| BX538083.1 | - | 6 | 804 | |||
| DB543411.1 | - | 1 | 111 | |||