siRNA Details
| Gene Symbol: KRT40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182497.3 | NP_872303.2 | 3 | 512 | ||
| XM_011524339.2 | XP_011522641.2 | 1 | 2279 | |||
| XM_017024189.1 | XP_016879678.1 | 1 | 2279 | |||
| GenBank mRNA | AJ786658.1 | - | 1 | 352 | ||
| AK093919.1 | - | 3 | 513 | |||
| BC130396.1 | - | 2 | 387 | 40146427 | ||
| BC144250.1 | - | 2 | 387 | 9052770 | ||
| BC150174.1 | - | 3 | 512 | 40147687 | ||
| BC150191.1 | - | 3 | 512 | 40148622 | ||

KRT40
Genomic Map