siRNA Details
| Gene Symbol: ZSWIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042697.1 | NP_001036162.1 | 4 | 378 | ||
| NM_001042698.1 | NP_001036163.1 | 4 | 378 | |||
| GenBank mRNA | AK093384.1 | - | 2 | 127 | ||
| BC018807.1 | - | 4 | 349 | |||
| BC044842.1 | - | 4 | 336 | |||
| BC066344.1 | - | 4 | 370 | |||
| BC089386.1 | - | 5 | 462 | |||
| BC094855.1 | - | 6 | 660 | |||
| BU075283.1 | - | 4 | 381 | |||
| DQ530600.1 | - | 4 | 280 | |||

ZSWIM7
Genomic Map