siRNA Details
| Gene Symbol: MIEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144900.1 | NP_001138372.1 | 3 | 411 | ||
| NM_139162.3 | NP_631901.2 | 3 | 411 | |||
| NM_148886.1 | NP_683684.2 | 3 | 259 | |||
| XM_017024190.1 | XP_016879679.1 | 3 | 756 | |||
| GenBank mRNA | AF467443.1 | - | 3 | 259 | ||
| AK056165.1 | - | 3 | 390 | |||
| AK128310.1 | - | 3 | 314 | |||
| BC014973.1 | - | 3 | 259 | 4859126 | ||
| DB183618.1 | - | 3 | 417 | |||

MIEF2
Genomic Map