siRNA Details
| Gene Symbol: SLC5A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042450.2 | NP_001035915.1 | 7, 8 | 684 | ||
| NM_001270649.1 | NP_001257578.1 | 7, 8 | 684 | |||
| NM_152351.4 | NP_689564.3 | 7, 8 | 684 | |||
| XM_011523652.1 | XP_011521954.1 | 7, 8 | 840 | |||
| XM_017024191.1 | XP_016879680.1 | 7, 8 | 841 | |||
| GenBank mRNA | AK057946.1 | - | 7, 8 | 685 | ||
| AK298345.1 | - | 7, 8 | 670 | |||
| BC034380.1 | - | 7, 8 | 629 | |||

SLC5A10
Genomic Map