siRNA Details
| Gene Symbol: ZNF816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031665.2 | NP_001026835.1 | 5 | 1399 | ||
| NM_001202456.1 | NP_001189385.1 | 4 | 1258 | |||
| NM_001202457.1 | NP_001189386.1 | 4 | 1248 | |||
| GenBank mRNA | AK291990.1 | - | 4 | 1162 | ||
| BC105739.1 | - | 4 | 1105 | 40026775 | ||
| BC105740.1 | - | 4 | 1108 | 40026776 | ||
| BC105930.1 | - | 4 | 1105 | 40026773 | ||

ZNF816
Genomic Map