siRNA Details
| Gene Symbol: ZNF816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031665.2 | NP_001026835.1 | 5 | 1820 | ||
| NM_001202456.1 | NP_001189385.1 | 4 | 1679 | |||
| NM_001202457.1 | NP_001189386.1 | 4 | 1669 | |||
| GenBank mRNA | AK291990.1 | - | 4 | 1583 | ||
| AL713804.1 | - | 1 | 332 | |||
| BC105739.1 | - | 4 | 1526 | 40026775 | ||
| BC105740.1 | - | 4 | 1529 | 40026776 | ||
| BC105930.1 | - | 4 | 1526 | 40026773 | ||

ZNF816
Genomic Map