siRNA Details
| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213598.3 | NP_998763.2 | 3 | 514 | ||
| GenBank mRNA | AK131547.1 | - | 3 | 502 | ||
| BC101012.2 | - | 3 | 157 | 40006636 | ||
| BC101013.1 | - | 3 | 157 | 40006637 | ||
| BC101015.1 | - | 3 | 157 | 40006640 | ||

ZNF543
Genomic Map