siRNA Details
| Gene Symbol: CEACAM20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102597.2 | NP_001096067.2 | 1, 2 | 76 | ||
| NM_001102598.2 | NP_001096068.2 | 1, 2 | 76 | |||
| NM_001102599.2 | NP_001096069.2 | 1, 2 | 76 | |||
| NM_001102600.2 | NP_001096070.2 | 1, 2 | 76 | |||
| GenBank mRNA | AY358129.1 | - | 1, 2 | 339 | ||
| EU008600.1 | - | 1, 2 | 76 | |||
| EU008601.1 | - | 1, 2 | 76 | |||
| EU008602.1 | - | 1, 2 | 76 | |||
| EU008603.1 | - | 1, 2 | 76 | |||

CEACAM20
Genomic Map