siRNA Details
| Gene Symbol: OR1M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004456.1 | NP_001004456.1 | 1 | 247 | ||
| GenBank mRNA | BC137551.1 | - | 1 | 313 | 9021551 | |
| BC137555.1 | - | 1 | 313 | 9021555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR1M1| Gene Symbol: OR1M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004456.1 | NP_001004456.1 | 1 | 247 | ||
| GenBank mRNA | BC137551.1 | - | 1 | 313 | 9021551 | |
| BC137555.1 | - | 1 | 313 | 9021555 | ||