siRNA Details
| Gene Symbol: ZNF813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004301.3 | NP_001004301.2 | 4 | 379 | ||
| GenBank mRNA | DB066632.1 | - | 3 | 252 | ||
| DB235327.1 | - | 4 | 370 | |||
| DR006226.1 | - | 2 | 199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF813| Gene Symbol: ZNF813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004301.3 | NP_001004301.2 | 4 | 379 | ||
| GenBank mRNA | DB066632.1 | - | 3 | 252 | ||
| DB235327.1 | - | 4 | 370 | |||
| DR006226.1 | - | 2 | 199 | |||