siRNA Details
| Gene Symbol: CCDC159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080503.2 | NP_001073972.2 | 7, 8 | 670 | ||
| XM_006722643.1 | XP_006722706.1 | 8, 9 | 721 | |||
| XM_011527678.2 | XP_011525980.2 | 7, 8 | 839 | |||
| XM_017026255.1 | XP_016881744.1 | 8, 9 | 721 | |||
| XM_017026256.1 | XP_016881745.1 | 8, 9 | 960 | |||
| XM_017026257.1 | XP_016881746.1 | 8, 9 | 784 | |||
| XM_017026258.1 | XP_016881747.1 | 7, 8 | 832 | |||
| XM_017026259.1 | XP_016881748.1 | 7, 8 | 830 | |||
| XM_017026260.1 | XP_016881749.1 | 7, 8 | 839 | |||
| XM_017026261.1 | XP_016881750.1 | 8, 9 | 962 | |||
| XM_017026262.1 | XP_016881751.1 | 7, 8 | 917 | |||
| XM_017026263.1 | XP_016881752.1 | 7, 8 | 910 | |||
| XM_017026264.1 | XP_016881753.1 | 8, 9 | 962 | |||
| GenBank mRNA | AK303641.1 | - | 7, 8 | 634 | ||
| BC038439.1 | - | 10, 11 | 1413 | |||

CCDC159
Genomic Map