siRNA Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270639.1 | NP_001257568.1 | 3 | 459 | ||
| NM_001270640.1 | NP_001257569.1 | 3 | 456 | |||
| NM_001270686.1 | NP_001257615.1 | 3 | 302 | |||
| NM_138334.3 | NP_612207.1 | 3 | 305 | |||
| XM_011526434.2 | XP_011524736.1 | 3 | 299 | |||
| GenBank mRNA | AF247787.1 | - | 2 | 234 | ||
| BC051380.1 | - | 3 | 263 | |||
| BC062416.1 | - | 3 | 282 | 6503623 | ||
| BP328576.1 | - | 3 | 302 | |||
| HY056543.1 | - | 3 | 460 | |||

JOSD2
Genomic Map