siRNA Details
| Gene Symbol: DRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001388.4 | NP_001379.1 | 2 | 370 | ||
| XM_005256499.2 | XP_005256556.1 | 2 | 352 | |||
| XM_011523704.1 | XP_011522006.1 | 2 | 352 | |||
| XM_011523705.1 | XP_011522007.1 | 2 | 352 | |||
| XM_011523706.1 | XP_011522008.1 | 2 | 265 | |||
| XM_017024296.1 | XP_016879785.1 | 2 | 265 | |||
| GenBank mRNA | AK295575.1 | - | 2 | 258 | ||
| AK313362.1 | - | 2 | 289 | |||
| BC000493.2 | - | 2 | 268 | 2820542 | ||
| BT006976.1 | - | 2 | 213 | |||
| X80754.1 | - | 2 | 260 | |||

DRG2
Genomic Map