siRNA Details
| Gene Symbol: NLRP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321057.1 | NP_001307986.1 | 5 | 1103 | ||
| NM_176810.2 | NP_789780.2 | 5 | 1103 | |||
| GenBank mRNA | AY154468.1 | - | 5 | 1078 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NLRP13| Gene Symbol: NLRP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321057.1 | NP_001307986.1 | 5 | 1103 | ||
| NM_176810.2 | NP_789780.2 | 5 | 1103 | |||
| GenBank mRNA | AY154468.1 | - | 5 | 1078 | ||