siRNA Details
| Gene Symbol: TMIGD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169126.1 | NP_001162597.1 | 2 | 420 | ||
| NM_144615.2 | NP_653216.2 | 2 | 420 | |||
| XM_017026284.1 | XP_016881773.1 | 2 | 428 | |||
| GenBank mRNA | AY358964.1 | - | 2 | 382 | ||
| BC015655.1 | - | 2 | 420 | 4811204 | ||

TMIGD2
Genomic Map