siRNA Details
| Gene Symbol: ZNF57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319083.1 | NP_001306012.1 | 3 | 243 | ||
| NM_173480.2 | NP_775751.1 | 3 | 342 | |||
| XM_011527682.2 | XP_011525984.1 | 2 | 210 | |||
| GenBank mRNA | AK302165.1 | - | 3 | 243 | ||
| AK314848.1 | - | 3 | 264 | |||
| BC028974.1 | - | 3 | 274 | 5138909 | ||
| BX537601.1 | - | 3 | 284 | |||
| EF534355.1 | - | 3 | 342 | |||

ZNF57
Genomic Map