siRNA Details
| Gene Symbol: IRGQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007561.2 | NP_001007562.1 | 3 | 1195 | ||
| XM_005258515.3 | XP_005258572.1 | 3 | 1274 | |||
| GenBank mRNA | AF322648.1 | - | 2 | 1225 | ||
| BC137020.1 | - | 2 | 1157 | 9021013 | ||
| BC137021.1 | - | 2 | 1157 | 9021014 | ||
| BC144451.1 | - | 2 | 1157 | 9052977 | ||

IRGQ
Genomic Map