siRNA Details
| Gene Symbol: ZNF428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182498.3 | NP_872304.2 | 3 | 610 | ||
| GenBank mRNA | AY257197.1 | - | 3 | 350 | ||
| BC015950.2 | - | 3 | 329 | 3908182 | ||
| BC045799.1 | - | 3 | 353 | 4813427 | ||
| BE901751.1 | - | 3 | 597 | |||
| BG121932.1 | - | 3 | 381 | |||
| CN355560.1 | - | 3 | 617 | |||

ZNF428
Genomic Map