siRNA Details
| Gene Symbol: MOB3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130807.2 | NP_570719.1 | 3, 4 | 747 | ||
| XM_011527683.2 | XP_011525985.1 | 3, 4 | 824 | |||
| GenBank mRNA | AJ580635.1 | - | 1, 2 | 588 | ||
| AK095471.1 | - | 3, 4 | 747 | |||
| AL832474.1 | - | 3, 4 | 783 | |||
| BC015049.1 | - | 2, 3 | 636 | 3919665 | ||

MOB3A
Genomic Map