siRNA Details
| Gene Symbol: GIPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133261.2 | NP_573568.1 | 6 | 1072 | ||
| XM_005259492.3 | XP_005259549.1 | 6 | 1125 | |||
| GenBank mRNA | AB073738.1 | - | 6 | 1072 | ||
| BX648927.1 | - | 1 | 18 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GIPC3| Gene Symbol: GIPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133261.2 | NP_573568.1 | 6 | 1072 | ||
| XM_005259492.3 | XP_005259549.1 | 6 | 1125 | |||
| GenBank mRNA | AB073738.1 | - | 6 | 1072 | ||
| BX648927.1 | - | 1 | 18 | |||