siRNA Details
| Gene Symbol: OR1I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004713.1 | NP_001004713.1 | 1 | 955 | ||
| GenBank mRNA | BC140930.1 | - | 1 | 1041 | 9056742 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR1I1| Gene Symbol: OR1I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004713.1 | NP_001004713.1 | 1 | 955 | ||
| GenBank mRNA | BC140930.1 | - | 1 | 1041 | 9056742 | |