siRNA Details
| Gene Symbol: RINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195833.1 | NP_001182762.1 | 8 | 1047 | ||
| NM_198445.3 | NP_940847.1 | 8 | 1031 | |||
| XM_006723015.2 | XP_006723078.1 | 8 | 1041 | |||
| XM_006723016.2 | XP_006723079.1 | 8 | 1009 | |||
| XM_011526454.2 | XP_011524756.1 | 8 | 1111 | |||
| XM_011526455.2 | XP_011524757.1 | 9 | 945 | |||
| GenBank mRNA | AK126119.1 | - | 5 | 1427 | ||
| AK127808.1 | - | 8 | 1006 | |||
| AK298327.1 | - | 8 | 1047 | |||

RINL
Genomic Map