siRNA Details
| Gene Symbol: LRRC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010847.1 | NP_001010847.1 | 2 | 995 | ||
| GenBank mRNA | BC016048.1 | - | 2 | 910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC38| Gene Symbol: LRRC38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010847.1 | NP_001010847.1 | 2 | 995 | ||
| GenBank mRNA | BC016048.1 | - | 2 | 910 | ||