siRNA Details
| Gene Symbol: KLHDC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007255.2 | NP_001007256.1 | 4 | 1067 | ||
| NM_152366.4 | NP_689579.3 | 4 | 1048 | |||
| NR_033385.1 | 5 | 817 | ||||
| NR_033386.1 | 5 | 836 | ||||
| GenBank mRNA | AK093268.1 | - | 1 | 1676 | ||
| AY601914.1 | - | 4 | 1066 | |||
| BC022077.1 | - | 4 | 1047 | 4822843 | ||
| BC032482.1 | - | 1 | 1705 | 5222141 | ||
| BC066896.1 | - | 4 | 1042 | 4828667 | ||

KLHDC9
Genomic Map