siRNA Details
| Gene Symbol: ATXN7L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153340.4 | NP_699171.3 | 6, 7 | 805 | ||
| XM_005270445.3 | XP_005270502.1 | 6, 7 | 936 | |||
| XM_005270446.2 | XP_005270503.1 | 6, 7 | 932 | |||
| XM_011540638.2 | XP_011538940.1 | 5, 6 | 835 | |||
| XM_011540639.2 | XP_011538941.1 | 6, 7 | 2205 | |||
| XM_011540641.2 | XP_011538943.1 | 6, 7 | 1538 | |||
| XM_011540644.2 | XP_011538946.1 | 8, 9 | 1020 | |||
| XM_011540646.2 | XP_011538948.1 | 6, 7 | 929 | |||
| XM_017000267.1 | XP_016855756.1 | 7, 8 | 1667 | |||
| XM_017000268.1 | XP_016855757.1 | 7, 8 | 1000 | |||
| XM_017000269.1 | XP_016855758.1 | 7, 8 | 1667 | |||
| XM_017000270.1 | XP_016855759.1 | 7, 8 | 1068 | |||
| GenBank mRNA | AK090460.1 | - | 4, 5 | 1653 | ||

ATXN7L2
Genomic Map